(Cyanomethyl)triphenylphosphonium chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL464937-25GEA
107
EUR
SIAL464937-25G
(Cyanomethyl)triphenylphosphonium chloride, Sigma-Aldrich®
(Cyanomethyl)triphenylphosphonium chloride
Formula:
C₂₀H₁₇ClNP MW: 337,79 g/mol |
UN:
3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...