Diphenylphosphoryl azide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Diphenylphosphonic azide, Diphenyl azidophosphate

M-Clarity™ quality level = MQ200

178756-100G 178756-25G 178756-5G
SIAL178756-100GEA 202 EUR
SIAL178756-100G SIAL178756-25G SIAL178756-5G
Diphenylphosphoryl azide, Sigma-Aldrich®
Diphenylphosphoryl azide
Formula: C₁₂H₁₀N₃O₃P
MW: 275.20 g/mol
MDL Number: MFCD00001987
CAS Number: 26386-88-9
EINECS: 247-644-0
UN: 3278
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR