6-O-Palmitoyl-L-ascorbic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate
M-Clarity™ quality level = MQ400
A1968-100G
A1968-1KG
A1968-25G
SIALA1968-100GEA
389
EUR
SIALA1968-100G
SIALA1968-1KG
SIALA1968-25G
6-O-Palmitoyl-L-ascorbic acid, Sigma-Aldrich®
6-O-Palmitoyl-L-ascorbic acid
Formula:
C₂₂H₃₈O₇ Boiling Pt: ∼250 °C Melting Pt: 107…117 °C |
MDL Number:
MFCD00005377 CAS Number: 137-66-6 EINECS: 205-305-4 |