4-Bromo-1-methyl-1H-pyrazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
696455-25G 696455-5G
SIAL696455-25GEA 121 EUR
SIAL696455-25G SIAL696455-5G
4-Bromo-1-methyl-1H-pyrazole, Sigma-Aldrich®
4-Bromo-1-methyl-1H-pyrazole
Formula: C₄H₅BrN₂
MW: 161.00 g/mol
MDL Number: MFCD02179565
CAS Number: 15803-02-8