1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,3-Dichloro-1,1,3,3-tetrakis(isopropyl)disiloxane
M-Clarity™ quality level = MQ200
337005-1G 337005-5G
SIAL337005-1GEA 42.5 EUR
SIAL337005-1G SIAL337005-5G
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, Sigma-Aldrich®
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Formula: C₁₂H₂₈Cl₂OSi₂
MW: 315.43 g/mol
MDL Number: MFCD00009655
CAS Number: 69304-37-6
UN: 1760
ADR: 8,II