Tin standard dissolved in oil, 5000 µg/g Sn in mineral oil for wear metal analysis, Specpure®
Supplier: Thermo Fisher Scientific
Specpure®
42450.18EA 110 EUR
42450.18 42450.1Y
Tin standard dissolved in oil, 5000 µg/g Sn in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Tin Standards
Physical Form: Liquid
MDL No.: MFCD00133862