(±)-Naringenin, Sigma-Aldrich®

Supplier: Merck

Synonyms: (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone, Naringenin

M-Clarity™ quality level = MQ300

W530098-500G W530098-SAMPLE
SIALW530098-500GEA 601 EUR
SIALW530098-500G SIALW530098-SAMPLE
(±)-Naringenin, Sigma-Aldrich®
(±)-Naringenin
Formula: C₁₅H₁₂O₅
MW: 272.25 g/mol
MDL Number: MFCD00006844
CAS Number: 67604-48-2
EINECS: 266-769-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR