N,N-Dimethyl 3-boronobenzenesulphonamide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(3-(N,N-Dimethylsulphamoyl)phenyl)boronic acid, 3-(Dimethylsulphamoyl)benzeneboronic acid
H52838.MD
H52838.03
H52838.MDEA
136
EUR
H52838.MD
H52838.03
N,N-Dimethyl 3-boronobenzenesulphonamide ≥98%
N,N-Dimethyl 3-boronobenzenesulphonamide
Purity: 98%
CAS No.: 871329-59-8
Molecular Formula: C8H12BNO4S
Formula Weight: 229.06
Melting Point: 88-92°
MDL No.: MFCD07363747
Formula:
C₈H₁₂BNO₄S Melting Pt: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363747 CAS Number: 871329-59-8 |