2-Hydroxy-4-methoxy-5-sulphobenzophenone hydrate tech. 85% may cont. up to 10% 2-propanol, technical
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid Hydrate
L12912.09 L12912.18
L12912.09EA 55.5 EUR
L12912.09 L12912.18
2-Hydroxy-4-methoxy-5-sulphobenzophenone hydrate tech. 85% may cont. up to 10% 2-propanol, technical
2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate
Purity: 85% CAS No.: 4065-45-6 Molecular Formula: C14H12O6S.xH2O Formula Weight: 308.31(anhy) Melting Point: 108-112° Beilstein Reference: 2889165 Merck Reference: 14,8983 MDL No.: MFCD00977409
Formula: C₁₄H₁₂O₆S·H₂O
MW: 308.31 g/mol (anhy)
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00024962
CAS Number: 4065-45-6
EINECS: 223-772-2
Merck Index: 14,8983