4-(Trifluoromethyl)benzoic anhydride ≥97%
Supplier: Thermo Fisher Scientific
Warning
H34480.03 H34480.06
H34480.03EA 33.8 EUR
H34480.03 H34480.06
4-(Trifluoromethyl)benzoic anhydride ≥97%
4-(Trifluoromethyl)benzoic anhydride
Purity: 97% CAS No.: 25753-16-6 Molecular Formula: C16H8F6O3 Formula Weight: 362.23 Melting Point: 127-131° MDL No.: MFCD00671577
Formula: C₁₆H₈F₆O₃
MW: 362.23 g/mol
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00671577
CAS Number: 25753-16-6