5-Chloro-2-benzoxazolinone 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 5-Chloro-2(3H)-benzoxazolone, 5-Chlorobenzo[d]oxazol-2(3H)-one, Chlorzoxazone, Chloroxazone
228951000 228950250
ACRO228951000EA 68.6 EUR
ACRO228951000 ACRO228950250
5-Chloro-2-benzoxazolinone 98%
5-Chloro-2-benzoxazolinone
Formula: C₇H₄ClNO₂
MW: 169.57 g/mol
Melting Pt: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00005717
CAS Number: 95-25-0
EINECS: 202-403-9
Order Now
Specification Test Results
Appearance White to yellow-brown Powder
Infrared spectrum Conforms
Melting point 189 to 194 °C
Water ≤1 % (K.F.)
Titration with NaOH ≥97.5 %