5-Chloro-2-benzoxazolinone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Chloro-2(3H)-benzoxazolone, 5-Chlorobenzo[d]oxazol-2(3H)-one, Chlorzoxazone, Chloroxazone
M-Clarity™ quality level = MQ100
C4397-100G
C4397-25G
SIALC4397-100GEA
238
EUR
SIALC4397-100G
SIALC4397-25G
5-Chloro-2-benzoxazolinone, Sigma-Aldrich®
5-Chloro-2-benzoxazolinone
Formula:
C₇H₄ClNO₂ MW: 169.57 g/mol |
MDL Number:
MFCD00005717 CAS Number: 95-25-0 EINECS: 202-403-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...