2-Bromoisonicotinamide ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Bromopyridine-4-carboxamide

L19854.03 L19854.06
L19854.03EA 73.1 EUR
L19854.03 L19854.06
2-Bromoisonicotinamide ≥97%
2-Bromoisonicotinamide

Purity: 97% CAS No.: 29840-73-1 Molecular Formula: C6H5BrN2O Formula Weight: 201.02 Melting Point: 192° Beilstein Reference: 471892

Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234156
CAS Number: 29840-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR