Benzyl-4-bromophenyl ketone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-(4-Bromophenyl)-2-phenylethanone
M-Clarity™ quality level = MQ100
515531-1G
515531-5G
SIAL515531-1GEA
44.8
EUR
SIAL515531-1G
SIAL515531-5G
Benzyl-4-bromophenyl ketone, Sigma-Aldrich®
Benzyl 4-bromophenyl ketone
Formula:
C₁₄H₁₁BrO MW: 275.14 g/mol |
MDL Number:
MFCD00016331 CAS Number: 2001-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...