4-(tert-Butyl)-2-chloroaniline

Supplier: Thermo Scientific

RF01508DA RF01508EA
MAYBRF01508DAEA 0 EUR
MAYBRF01508DA MAYBRF01508EA
4-(tert-Butyl)-2-chloroaniline
4-(tert-Butyl)-2-chloroaniline
Formula: C₁₀H₁₄ClN
MDL Number: MFCD00052272
CAS Number: 42265-67-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR