Erucic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(Z)-Docos-13-enoic acid
M-Clarity™ quality level = MQ200
E3385-1G
E3385-5G
SIALE3385-1GEA
202
EUR
SIALE3385-1G
SIALE3385-5G
Erucic acid, Sigma-Aldrich®
Erucic acid
Formula:
C₂₂H₄₂O₂ MW: 338.58 g/mol |
MDL Number:
MFCD00063188 CAS Number: 112-86-7 EINECS: 204-011-3 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...