2',4'-Dihydroxy-3'-propylacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-(2,4-Dihydroxy-3-propylphenyl)ethanone
426903-25G
426903-5G
SIAL426903-25GEA
319
EUR
SIAL426903-25G
SIAL426903-5G
2',4'-Dihydroxy-3'-propylacetophenone, Sigma-Aldrich®
2',4'-Dihydroxy-3'-propylacetophenone
Formula:
C₁₁H₁₄O₃ MW: 194.2300 g/mol |
MDL Number:
MFCD00068616 CAS Number: 40786-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...