Potassium acetate 5 mol/l (5 N) in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid potassium salt
95843-100ML-F 95843-250ML-F
SIAL95843-100ML-FEA 150 EUR
SIAL95843-100ML-F SIAL95843-250ML-F
Potassium acetate 5 mol/l (5 N) in aqueous solution, Sigma-Aldrich®
Potassium acetate
Formula: CH₃COOK
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM