Potassium acetate 8 M in aqueous solution
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Acetic acid potassium salt
J63372.AE J63372.AK
J63372.AEEA 56.9 EUR
J63372.AE J63372.AK
Potassium acetate 8 M in aqueous solution
Potassium acetate
CAS No.: 127-08-2 Molecular Formula: C2H3KO2 Formula Weight: 98.15 Physical Form: Liquid MDL No.: MFCD00012458
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM