1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
Supplier: Thermo Scientific
CD03278.1
CD03278.10
MAYBCD03278.1EA
0
EUR
MAYBCD03278.1
MAYBCD03278.10
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
Formula:
C₁₂H₇F₆N MW: 279.19 g/mol |
MDL Number:
MFCD00053048 CAS Number: 175136-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...