D-(+)-Fucose, Sigma-Aldrich®

Supplier: Merck

Synonyms: alpha-D-Fucose

M-Clarity™ quality level = MQ200

F8150-1G F8150-500MG F8150-5G
SIALF8150-1GEA 141 EUR
SIALF8150-1G SIALF8150-500MG SIALF8150-5G
D-(+)-Fucose, Sigma-Aldrich®
D(+)-Fucose
Formula: C₆H₁₂O₅
MDL Number: MFCD00135603
CAS Number: 3615-37-0
EINECS: 222-792-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR