(5-Carboxypentyl)triphenylphosphonium bromide
Supplier: Thermo Scientific
KM08684.1 KM08684EA KM08684.50
MAYBKM08684.1EA 0 EUR
MAYBKM08684.1 MAYBKM08684EA MAYBKM08684.50
(5-Carboxypentyl)triphenylphosphonium bromide
(5-Carboxypentyl)triphenylphosphonium bromide
Formula: C₂₄H₂₆BrO₂P
MW: 457.36 g/mol
MDL Number: MFCD00055556
CAS Number: 50889-29-7