4-Amino-3-chlorobenzonitrile

Supplier: Thermo Scientific

SEW01283.1 SEW01283EA
MAYBSEW01283.1EA 0 EUR
MAYBSEW01283.1 MAYBSEW01283EA
4-Amino-3-chlorobenzonitrile
4-Amino-3-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00052917
CAS Number: 21803-75-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR