Barium isopropoxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Barium i-propoxide

M-Clarity™ quality level = MQ100

449679-10G 449679-2G
SIAL449679-10GEA 318 EUR
SIAL449679-10G SIAL449679-2G
Barium isopropoxide, Sigma-Aldrich®
Barium isopropoxide
Formula: C₆H₁₄BaO₂
MW: 255.501 g/mol
MDL Number: MFCD00050486
CAS Number: 24363-37-9
UN: 3205
ADR: 4.2,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR