Sodium bis(trimethylsilyl)amide 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Synonyms: HMDS sodium salt, N-Sodiohexamethyldisilazane, Sodium bis(trimethylsilyl)amide, HMDS-Na
M-Clarity™ quality level = MQ200
245585-100ML 245585-800ML
SIAL245585-100MLEA 58.5 EUR
SIAL245585-100ML SIAL245585-800ML
Sodium bis(trimethylsilyl)amide 1 mol/l in THF, Sigma-Aldrich®
Sodium bis(trimethylsilyl)amide
Formula: C₆H₁₈NNaSi₂
MW: 183.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009835
CAS Number: 1070-89-9
EINECS: 213-983-8
UN: 2924
ADR: 3,II