4-tert-Butylaniline
Supplier: Thermo Scientific
SB01038DA
SB01038.10
SB01038.50
MAYBSB01038DAEA
0
EUR
MAYBSB01038DA
MAYBSB01038.10
MAYBSB01038.50
4-tert-Butylaniline
4-tert-Butylaniline
Formula:
C₁₀H₁₅N Boiling Pt: 238 °C (1013 hPa) Melting Pt: 15 °C Density: 0.941 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007899 CAS Number: 769-92-6 EINECS: 212-215-9 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...