Mineral oil, white (petroleum), 75 cSt
Supplier: Thermo Scientific
Danger
14621.MQEA 46.7 EUR
14621.MQ 14621.X2
Mineral oil, white (petroleum), 75 cSt
Mineral oil, white (petroleum)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-7