Magnetic stirring bars, cylindrical, PTFE
Supplier: SciLabware
SWN652 SWN654 SWN656 SWN658 SWN660 SWN662 SWN664 SWN666 SWN668 SWN670 SWN672 SWN674 SWN676
442-1176EA 12.2 EUR
442-1176 442-1177 442-1167 442-1168 442-1169 442-1170 442-1171 442-1172 442-1173 442-1174 442-1175 442-1160 442-1161
Magnetic stirring bars, cylindrical, PTFE
Stirrers Magnetic Stirring Bars
PTFE covered, plain design with rounded ends, suitable for use in a wide range of flat bottomed containers.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)