(S)-(+)-1-Boc-3-hydroxypyrrolidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

M-Clarity™ quality level = MQ100

634786-5G 634786-1G
SIAL634786-5GEA 123 EUR
SIAL634786-5G SIAL634786-1G
(S)-(+)-1-Boc-3-hydroxypyrrolidine, Sigma-Aldrich®
(S)-(+)-1-Boc-3-hydroxypyrrolidine
Formula: C₉H₁₇NO₃
MW: 187.2362 g/mol
MDL Number: MFCD01317839
CAS Number: 101469-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR