Estradiol, Sigma-Aldrich®

Supplier: Merck

Synonyms: beta-Estradiol, 17β-Estradiol

MQ200

E8875-1G E8875-100G E8875-250MG E8875-25G E8875-5G
SIALE8875-1GEA 89.7 EUR
SIALE8875-1G SIALE8875-100G SIALE8875-250MG SIALE8875-25G SIALE8875-5G
Estradiol, Sigma-Aldrich®
Estradiol
Formula: C₁₈H₂₄O₂
MW: 272.39 g/mol
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR