Estradiol, Sigma-Aldrich®

Supplier: Merck

Synonyms: beta-Estradiol, 17β-Estradiol

M-Clarity™ quality level = MQ200

E1132-1VL E1132-5VL
SIALE1132-1VLEA 110 EUR
SIALE1132-1VL SIALE1132-5VL
Estradiol, Sigma-Aldrich®
Estradiol
Formula: C₁₈H₂₄O₂
MW: 272.39 g/mol
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR