D-(+)-Arabitol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
M-Clarity™ quality level = MQ200
A3381-100G
A3381-25G
SIALA3381-100GEA
1080
EUR
SIALA3381-100G
SIALA3381-25G
D-(+)-Arabitol, Sigma-Aldrich®
D(+)-Arabitol
Formula:
C₅H₁₂O₅ MW: 152.15 g/mol |
MDL Number:
MFCD00004709 CAS Number: 488-82-4 EINECS: 207-686-2 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...