Brefeldin A ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
J62340.MB
J62340.MF
J62340.MBEA
382
EUR
J62340.MB
J62340.MF
Brefeldin A ≥99%
Brefeldin A
Formula:
C₁₆H₂₄O₄ Melting Pt: 201…205 °C Storage Temperature: Freezer |
MDL Number:
MFCD00083258 CAS Number: 20350-15-6 Merck Index: 14,1369 |