Ethylamine 2 mol/l in methanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Monoethylamine, Aminoethane, Ethanamine
M-Clarity™ quality level = MQ100
395064-100ML 395064-800ML
SIAL395064-100MLEA 109 EUR
SIAL395064-100ML SIAL395064-800ML
Ethylamine 2 mol/l in methanol, Sigma-Aldrich®
Ethylamine
Formula: C₂H₇N
MW: 45,08 g/mol
MDL Number: MFCD00008160
CAS Number: 75-04-7
EINECS: 200-834-7