Ethylamine 2 mol/l in THF, AcroSeal®
Supplier: Thermo Scientific
Danger
Synonyms: Monoethylamine, Aminoethane, Ethanamine
370251000 370258000
370251000.EA 72.5 EUR
370251000. 370258000.
Ethylamine 2 mol/l in THF, AcroSeal®
Ethylamine
Formula: C₂H₇N
MW: 45,08 g/mol
Density: 0.861 g/cm³
Flash Pt: –34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008160
CAS Number: 75-04-7
EINECS: 200-834-7
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Strength 1.90 to 2.20 M