Ethylamine 2 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Synonyms: Monoethylamine, Aminoethane, Ethanamine
M-Clarity™ quality level = MQ100
395072-100ML 395072-800ML
SIAL395072-100MLEA 91.5 EUR
SIAL395072-100ML SIAL395072-800ML
Ethylamine 2 mol/l in THF, Sigma-Aldrich®
Ethylamine
Formula: C₂H₇N
MW: 45,08 g/mol
MDL Number: MFCD00008160
CAS Number: 75-04-7
EINECS: 200-834-7