Diethyl cyclopropane-1,1-dicarboxylate
Supplier: Thermo Scientific
Warning
Synonyms: Diethyl 1,1-cyclopropanedicarboxylate, Diethyl 1,1-cyclopropane dicarboxyliate
H56568.03 H56568.06 H56568.14
H56568.03EA 37.6 EUR
H56568.03 H56568.06 H56568.14
Diethyl cyclopropane-1,1-dicarboxylate
Diethyl cyclopropane-1,1-dicarboxylate
Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Boiling Pt: 94…96 °C (10 mmHg)
Density: 1.055
Flash Pt: 87 °C (189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001283
CAS Number: 1559-02-0
EINECS: 216-321-6