1-(3,4-Dichlorobenzyl)piperazine ≥97%
Supplier: Thermo Fisher Scientific
Warning
H55685.06 H55685.14
H55685.06EA 239 EUR
H55685.06 H55685.14
1-(3,4-Dichlorobenzyl)piperazine ≥97%
1-(3,4-Dichlorobenzyl)piperazine
Formula: C₁₁H₁₄Cl₂N₂
Boiling Pt: 124…128 °C (0.02 mmHg)
Density: 1.225
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03407488
CAS Number: 55513-17-2