1-Bromoisoquinoline 95+%
Supplier: Thermo Fisher Scientific
Danger
392590050 392590010
ACRO392590050EA 336 EUR
ACRO392590050 ACRO392590010
1-Bromoisoquinoline 95+%
1-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.0546 g/mol
Melting Pt: 43…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00234478
CAS Number: 1532-71-4
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Yellow to brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 42 to 48 °C
HPLC ≥95.0 %