3-Bromoimidazo[1,2-a]pyridine

Supplier: Thermo Scientific

CC25410.250 CC25410DA
MAYBCC25410.250EA 0 EUR
MAYBCC25410.250 MAYBCC25410DA
3-Bromoimidazo[1,2-a]pyridine
3-Bromoimidazo[1,2-a]pyridine
Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD00014556
CAS Number: 4926-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR