2-Methylimidazo[1,2-a]pyridine 95%
Supplier: Thermo Scientific
Warning
H35114.MD H35114.03
H35114.MDEA 0 EUR
H35114.MD H35114.03
2-Methylimidazo[1,2-a]pyridine 95%
2-Methylimidazo[1,2-a]pyridine
Formula: C₈H₈N₂
MW: 132.17 g/mol
Boiling Pt: 148 °C (20 mmHg)
Melting Pt: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD02251330
CAS Number: 934-37-2