1H-Benzimidazole-2-sulphonic acid

Supplier: Thermo Scientific

Synonyms: 1H-Benzo[d]imidazole-2-sulphonic acid

BTB12616.1 BTB12616.10 BTB12616.50
MAYBBTB12616.1EA 0 EUR
MAYBBTB12616.1 MAYBBTB12616.10 MAYBBTB12616.50
1H-Benzimidazole-2-sulphonic acid
1H-Benzimidazole-2-sulphonic acid
Formula: C₇H₆N₂O₃S
MDL Number: MFCD00100972
CAS Number: 40828-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR