2-Butyl-5-chloroimidazole-4-carbaldehyde
Supplier: Thermo Scientific
Synonyms:
2-Butyl-4-chloro-5-formylimidazole, 2-(But-1-yl)-5-chloro-1H-imidazole-4-carboxaldehyde, 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde
SB02042.1
SB02042.10
SB02042.50
MAYBSB02042.1EA
0
EUR
MAYBSB02042.1
MAYBSB02042.10
MAYBSB02042.50
2-Butyl-5-chloroimidazole-4-carbaldehyde
2-Butyl-5-chloroimidazole-4-carbaldehyde
Formula:
C₈H₁₁ClN₂O MW: 186.6387 g/mol |
MDL Number:
MFCD01934396 CAS Number: 83857-96-9 EINECS: 410-260-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...