N-(2,4-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide
Supplier: Thermo Scientific
RF04574.250
RF04574.1
MAYBRF04574.250EA
0
EUR
MAYBRF04574.250
MAYBRF04574.1
N-(2,4-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide
N-(2,4-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide
Formula:
C₂₂H₁₆F₁₃NO₃ |
MDL Number:
MFCD00179736 CAS Number: 288161-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...