Peppermint oil, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W284815-1KG-K W284815-9KG-K
SIALW284815-1KG-KEA 246 EUR
SIALW284815-1KG-K SIALW284815-9KG-K
Peppermint oil, Sigma-Aldrich®
Peppermint oil
MDL Number: MFCD00147870
CAS Number: 8006-90-4