Peppermint oil, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W284823-1KG-K W284823-4KG-K
SIALW284823-1KG-KEA 231 EUR
SIALW284823-1KG-K SIALW284823-4KG-K
Peppermint oil, Sigma-Aldrich®
Peppermint oil
MDL Number: MFCD00147870
CAS Number: 8006-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR