2-Chloro-4-(methylsulphonyl)benzoic acid

Supplier: Thermo Scientific

Synonyms: 2-Chloro-4-methylsulfonylbenzoic acid, 2-Chloro-4-methylsulphonylbenzoic acid

JFD01500.1 JFD01500EA JFD01500.50
MAYBJFD01500.1EA 0 EUR
MAYBJFD01500.1 MAYBJFD01500EA MAYBJFD01500.50
2-Chloro-4-(methylsulphonyl)benzoic acid
2-Chloro-4-(methylsulphonyl)benzoic acid
Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
MDL Number: MFCD00216496
CAS Number: 53250-83-2
EINECS: 406-520-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR