Benzhydryl isothiocyanate

Supplier: Thermo Scientific

S03388.1 S03388EA
MAYBS03388.1EA 0 EUR
MAYBS03388.1 MAYBS03388EA
Benzhydryl isothiocyanate
Benzhydryl isothiocyanate
Formula: C₁₄H₁₁NS
MDL Number: MFCD00046815
CAS Number: 3550-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR