3-Mercaptobenzoic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

451436-1G 451436-5G
SIAL451436-1GEA 85.3 EUR
SIAL451436-1G SIAL451436-5G
3-Mercaptobenzoic acid, Sigma-Aldrich®
3-Mercaptobenzoic acid
Formula: C₇H₆O₂S
MW: 154.1863 g/mol
MDL Number: MFCD00238636
CAS Number: 4869-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR