Clamps
Supplier: SEMADENI
1076 1078 1080 1081 1082 1084 1075 1077
SEMA1076EA 8.8 EUR
SEMA1076 SEMA1078 SEMA1080 SEMA1081 SEMA1082 SEMA1084 SEMA1075 SEMA1077
Clamps
Clamps General Purpose Clamps
Clamps made of polyacetal (POM), in light blue.
  • Chemical and temperature resistant
  • Sticks (80 mm high) can be cut to the desired height